Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGATCTCCTCTTGTGCTCTTCCA[A/G]TTTCCAAGGATGTTCCAACTTGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 173445 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PECAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PECAM1 - platelet and endothelial cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000442.4 | 2912 | UTR 3 | NP_000433.4 | |||
XM_005276880.1 | 2912 | UTR 3 | XP_005276937.1 | |||
XM_005276881.1 | 2912 | UTR 3 | XP_005276938.1 | |||
XM_005276882.1 | 2912 | UTR 3 | XP_005276939.1 | |||
XM_005276883.2 | 2912 | UTR 3 | XP_005276940.1 | |||
XM_011524889.2 | 2912 | UTR 3 | XP_011523191.1 | |||
XM_011524890.1 | 2912 | UTR 3 | XP_011523192.1 | |||
XM_017024738.1 | 2912 | UTR 3 | XP_016880227.1 | |||
XM_017024739.1 | 2912 | UTR 3 | XP_016880228.1 | |||
XM_017024740.1 | 2912 | UTR 3 | XP_016880229.1 | |||
XM_017024741.1 | 2912 | UTR 3 | XP_016880230.1 |