Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606357 MIM: 610373 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX21 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DDX21 - DEAD-box helicase 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256910.1 | Intron | NP_001243839.1 | ||||
NM_004728.3 | Intron | NP_004719.2 | ||||
XM_011540336.1 | Intron | XP_011538638.1 | ||||
XM_017016910.1 | Intron | XP_016872399.1 |
DDX50 - DEAD-box helicase 50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |