Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
52 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180902 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RYR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RYR2 - ryanodine receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035.2 | Intron | NP_001026.2 | ||||
XM_006711802.3 | Intron | XP_006711865.1 | ||||
XM_006711803.3 | Intron | XP_006711866.1 | ||||
XM_006711804.3 | Intron | XP_006711867.1 | ||||
XM_006711805.3 | Intron | XP_006711868.1 | ||||
XM_006711806.3 | Intron | XP_006711869.1 | ||||
XM_006711807.3 | Intron | XP_006711870.1 | ||||
XM_006711808.3 | Intron | XP_006711871.1 | ||||
XM_006711809.3 | Intron | XP_006711872.1 | ||||
XM_006711810.3 | Intron | XP_006711873.1 | ||||
XM_017002028.1 | Intron | XP_016857517.1 |
Set Membership: |
HapMap Validated |