Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCCTCCCATTCCTGACAATTTC[A/G]TGGGGTCCTTGATATCTCCCCTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609373 MIM: 609347 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM3B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KDM3B - lysine demethylase 3B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REEP2 - receptor accessory protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271803.1 | Intron | NP_001258732.1 | ||||
NM_016606.3 | Intron | NP_057690.2 |
Set Membership: |
HapMap |