Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610469 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP3M2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AP3M2 - adaptor related protein complex 3 mu 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134296.1 | 210 | Intron | NP_001127768.1 | |||
NM_006803.3 | 210 | Intron | NP_006794.1 | |||
XM_017012977.1 | 210 | Intron | XP_016868466.1 | |||
XM_017012978.1 | 210 | Intron | XP_016868467.1 | |||
XM_017012979.1 | 210 | UTR 5 | XP_016868468.1 |
Set Membership: |
HapMap |