Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCAGCGCTTGAGAAGGTGAGAC[C/G]TGGGAGCTGAGGCTCCCGGAGGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615519 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DRC3 - dynein regulatory complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130090.1 | 202 | Intron | NP_001123562.1 | |||
NM_001130091.1 | 202 | Intron | NP_001123563.1 | |||
NM_001130092.1 | 202 | Intron | NP_001123564.1 | |||
NM_031294.3 | 202 | Intron | NP_112584.3 | |||
XM_006721583.3 | 202 | Intron | XP_006721646.1 | |||
XM_011524018.2 | 202 | Intron | XP_011522320.1 | |||
XM_011524019.2 | 202 | Intron | XP_011522321.1 | |||
XM_011524020.2 | 202 | UTR 5 | XP_011522322.1 | |||
XM_011524021.2 | 202 | Intron | XP_011522323.1 | |||
XM_011524022.2 | 202 | Intron | XP_011522324.1 | |||
XM_011524023.2 | 202 | Intron | XP_011522325.1 | |||
XM_011524025.2 | 202 | Intron | XP_011522327.1 | |||
XM_011524026.2 | 202 | Intron | XP_011522328.1 |
TOM1L2 - target of myb1 like 2 membrane trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033551.2 | 202 | Intron | NP_001028723.1 | |||
NM_001082968.1 | 202 | Intron | NP_001076437.1 | |||
NM_001288786.1 | 202 | Intron | NP_001275715.1 | |||
NM_001288787.1 | 202 | Intron | NP_001275716.1 | |||
NM_001288788.1 | 202 | Intron | NP_001275717.1 | |||
NM_001288789.1 | 202 | Intron | NP_001275718.1 | |||
XM_005256461.2 | 202 | Intron | XP_005256518.1 | |||
XM_005256462.1 | 202 | Intron | XP_005256519.1 | |||
XM_005256463.2 | 202 | Intron | XP_005256520.1 | |||
XM_005256466.1 | 202 | Intron | XP_005256523.1 | |||
XM_011523662.1 | 202 | Intron | XP_011521964.1 | |||
XM_017024201.1 | 202 | Intron | XP_016879690.1 |