Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGGAGAGGCCATTCAAGCACATG[G/T]GGAGGGTGGGAAGTGAGAGATAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604212 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PARN - poly(A)-specific ribonuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134477.2 | Intron | NP_001127949.1 | ||||
NM_001242992.1 | Intron | NP_001229921.1 | ||||
NM_002582.3 | Intron | NP_002573.1 | ||||
XM_011522510.2 | Intron | XP_011520812.1 | ||||
XM_011522511.2 | Intron | XP_011520813.1 | ||||
XM_011522513.2 | Intron | XP_011520815.1 | ||||
XM_011522514.1 | Intron | XP_011520816.1 | ||||
XM_017023258.1 | Intron | XP_016878747.1 | ||||
XM_017023259.1 | Intron | XP_016878748.1 | ||||
XM_017023260.1 | Intron | XP_016878749.1 |
Set Membership: |
HapMap |