Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATTTGCTGTGGACTTTATTTCAA[A/C]TTTCGTATTTCAAGTATTTCTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610589 MIM: 173120 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP11A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ARHGAP11A - Rho GTPase activating protein 11A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCG5 - secretogranin V | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |