Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTGGACAATATGCCAGATGTGG[G/A]GGAGGAGGCATAGGGGGGCTGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
49 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616088 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf186 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C1orf186 - chromosome 1 open reading frame 186 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007544.3 | 1392 | Intron | NP_001007545.1 | |||
XM_005272738.3 | 1392 | Intron | XP_005272795.1 |
LOC100505650 - uncharacterized LOC100505650 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017003088.1 | 1392 | UTR 3 | XP_016858577.1 |