Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTGGGCAACGTAAGGAGACCCC[A/G]TGTCTATTTTCATTAAAAAAAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154580 MIM: 607776 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAN2C1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAN2C1 - mannosidase alpha class 2C member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIN3A - SIN3 transcription regulator family member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145357.1 | Intron | NP_001138829.1 | ||||
NM_001145358.1 | Intron | NP_001138830.1 | ||||
NM_015477.2 | Intron | NP_056292.1 | ||||
XM_006720465.3 | Intron | XP_006720528.1 | ||||
XM_006720466.3 | Intron | XP_006720529.1 | ||||
XM_006720467.3 | Intron | XP_006720530.1 |
Set Membership: |
HapMap |