Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604750 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF226 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZNF226 - zinc finger protein 226 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032372.1 | 35 | UTR 5 | NP_001027544.1 | |||
NM_001032373.1 | 35 | Intron | NP_001027545.1 | |||
NM_001032374.1 | 35 | Intron | NP_001027546.1 | |||
NM_001146220.2 | 35 | Intron | NP_001139692.1 | |||
NM_001319088.1 | 35 | Intron | NP_001306017.1 | |||
NM_001319089.1 | 35 | Intron | NP_001306018.1 | |||
NM_001319090.1 | 35 | UTR 5 | NP_001306019.1 | |||
NM_015919.3 | 35 | Intron | NP_057003.2 | |||
NM_016444.2 | 35 | UTR 5 | NP_057528.2 | |||
XM_005259227.2 | 35 | Intron | XP_005259284.1 | |||
XM_006723367.3 | 35 | Intron | XP_006723430.1 | |||
XM_006723368.3 | 35 | Intron | XP_006723431.1 | |||
XM_006723369.2 | 35 | Intron | XP_006723432.1 | |||
XM_017027262.1 | 35 | UTR 5 | XP_016882751.1 | |||
XM_017027263.1 | 35 | Intron | XP_016882752.1 | |||
XM_017027264.1 | 35 | Intron | XP_016882753.1 | |||
XM_017027265.1 | 35 | UTR 5 | XP_016882754.1 |
ZNF234 - zinc finger protein 234 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |