Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGGGTTGAGGGTGGGGGTGCAG[A/G]GAGGAAGAACACCTGTGTGCAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612660 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFX7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RFX7 - regulatory factor X7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022841.5 | 2335 | Intron | NP_073752.5 | |||
XM_005254603.3 | 2335 | Intron | XP_005254660.2 | |||
XM_011521925.2 | 2335 | Intron | XP_011520227.1 | |||
XM_017022506.1 | 2335 | UTR 5 | XP_016877995.1 | |||
XM_017022507.1 | 2335 | Intron | XP_016877996.1 | |||
XM_017022508.1 | 2335 | Intron | XP_016877997.1 |
TEX9 - testis expressed 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286449.1 | 2335 | Intron | NP_001273378.1 | |||
NM_198524.2 | 2335 | Intron | NP_940926.1 | |||
XM_005254359.4 | 2335 | Intron | XP_005254416.1 | |||
XM_005254361.3 | 2335 | Intron | XP_005254418.1 | |||
XM_011521530.2 | 2335 | Intron | XP_011519832.1 | |||
XM_017022160.1 | 2335 | Intron | XP_016877649.1 | |||
XM_017022161.1 | 2335 | Intron | XP_016877650.1 | |||
XM_017022162.1 | 2335 | Intron | XP_016877651.1 | |||
XM_017022163.1 | 2335 | Intron | XP_016877652.1 | |||
XM_017022164.1 | 2335 | Intron | XP_016877653.1 | |||
XM_017022166.1 | 2335 | Intron | XP_016877655.1 | |||
XM_017022167.1 | 2335 | Intron | XP_016877656.1 |