Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCACTGCCTTCTAGAAGGGCAAA[C/T]ACAGTGTCAAATACGCTGGGTAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300375 MIM: 300368 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHST7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHST7 - carbohydrate sulfotransferase 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC9A7 - solute carrier family 9 member A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257291.1 | Intron | NP_001244220.1 | ||||
NM_032591.2 | Intron | NP_115980.1 | ||||
XM_005272681.4 | Intron | XP_005272738.1 | ||||
XM_005272682.4 | Intron | XP_005272739.1 | ||||
XM_006724564.3 | Intron | XP_006724627.1 | ||||
XM_011543990.2 | Intron | XP_011542292.1 | ||||
XM_017029905.1 | Intron | XP_016885394.1 | ||||
XM_017029906.1 | Intron | XP_016885395.1 |