Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGGTTAAGATGGTACATTTTAC[C/G]TGATGTGTTTTTTACCACAATAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604309 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM180A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM180A - family with sequence similarity 180 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_205855.3 | 1401 | UTR 3 | NP_995327.1 | |||
XM_006715983.3 | 1401 | UTR 3 | XP_006716046.1 |
SLC13A4 - solute carrier family 13 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |