Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAAGTTTGGGGTAACAGTCTTTC[A/G]TGAGTTACTTGATAGAGGCTATTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613514 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100130331 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100130331 - POTE ankyrin domain family, member F pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZP4 - zona pellucida glycoprotein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021186.4 | Intron | NP_067009.1 |