Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCCCAACTCAAACATTTTTCTGT[A/G]TGATTTATGTTATACCTGGGCATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAMTOR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LAMTOR4 - late endosomal/lysosomal adaptor, MAPK and MTOR activator 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008395.3 | Intron | NP_001008396.1 | ||||
NM_001318236.1 | Intron | NP_001305165.1 | ||||
NM_001318237.1 | Intron | NP_001305166.1 | ||||
XM_017012198.1 | Intron | XP_016867687.1 | ||||
XM_017012199.1 | Intron | XP_016867688.1 |
MBLAC1 - metallo-beta-lactamase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |