Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACCACAGTCTGGGGAGAGGAGC[C/T]GGGGAAGTGAGAAGTGTTCTGGAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172470 MIM: 607940 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHKG1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
PHKG1 - phosphorylase kinase catalytic subunit gamma 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258459.1 | 85 | Intron | NP_001245388.1 | |||
NM_001258460.1 | 85 | Intron | NP_001245389.1 | |||
NM_006213.4 | 85 | Intron | NP_006204.1 | |||
XM_005271772.4 | 85 | Intron | XP_005271829.1 | |||
XM_017012324.1 | 85 | Intron | XP_016867813.1 | |||
XM_017012325.1 | 85 | Intron | XP_016867814.1 | |||
XM_017012326.1 | 85 | Intron | XP_016867815.1 | |||
XM_017012327.1 | 85 | UTR 5 | XP_016867816.1 |
SUMF2 - sulfatase modifying factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |