Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTCCTTTTCCCCAGGCCCACCC[C/G]TGAACGTGAACATCACCAGCCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176889 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPN7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PTPN7 - protein tyrosine phosphatase, non-receptor type 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTPRVP - protein tyrosine phosphatase, receptor type V, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |