Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC35G2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC35G2 - solute carrier family 35 member G2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001097599.1 | 520 | Intron | NP_001091068.1 | |||
NM_001097600.1 | 520 | Intron | NP_001091069.1 | |||
NM_025246.2 | 520 | UTR 5 | NP_079522.2 | |||
XM_006713773.3 | 520 | Intron | XP_006713836.1 | |||
XM_011513214.2 | 520 | Intron | XP_011511516.1 | |||
XM_017007289.1 | 520 | Intron | XP_016862778.1 | |||
XM_017007290.1 | 520 | Intron | XP_016862779.1 | |||
XM_017007291.1 | 520 | Intron | XP_016862780.1 |
Set Membership: |
HapMap |