Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607724 MIM: 610395 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GLIPR1L1 - GLI pathogenesis related 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304964.1 | 2272 | Intron | NP_001291893.1 | |||
NM_152779.3 | 2272 | Intron | NP_689992.1 | |||
XM_011538134.2 | 2272 | UTR 5 | XP_011536436.2 | |||
XM_017019134.1 | 2272 | UTR 5 | XP_016874623.1 |
Set Membership: |
HapMap Validated |