Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCCAGCCAAAAGCTTTAAATCA[C/T]TGGTGATTATTACAAGCTGCAACTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614184 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIS3L2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DIS3L2 - DIS3 like 3'-5' exoribonuclease 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257281.1 | Intron | NP_001244210.1 | ||||
NM_001257282.1 | Intron | NP_001244211.1 | ||||
NM_152383.4 | Intron | NP_689596.4 |
Set Membership: |
HapMap |