Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603135 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CUL2 - cullin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198777.1 | Intron | NP_001185706.1 | ||||
NM_001198778.1 | Intron | NP_001185707.1 | ||||
NM_001198779.1 | Intron | NP_001185708.1 | ||||
NM_001324375.1 | Intron | NP_001311304.1 | ||||
NM_001324376.1 | Intron | NP_001311305.1 | ||||
NM_003591.3 | Intron | NP_003582.2 | ||||
XM_011519743.1 | Intron | XP_011518045.1 | ||||
XM_011519744.1 | Intron | XP_011518046.1 | ||||
XM_011519745.1 | Intron | XP_011518047.1 | ||||
XM_011519747.1 | Intron | XP_011518049.1 |
Set Membership: |
HapMap |