Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTTTTATTTGTTGGACAGTTAAT[A/G]TTTGGATTTTGTTTTTCATTTAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605184 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PAIP1 - poly(A) binding protein interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006451.4 | Intron | NP_006442.2 | ||||
NM_182789.3 | Intron | NP_877590.1 | ||||
NM_183323.2 | Intron | NP_899152.1 | ||||
XM_005248230.3 | Intron | XP_005248287.1 | ||||
XM_017008956.1 | Intron | XP_016864445.1 |
Set Membership: |
Validated |