Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGCTGCTTGGTTTGGTGGCTCA[A/G]AGACATCATCCAGGTCTCAGGGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194536 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF12 - zinc finger protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006956.2 | 4154 | UTR 3 | NP_008887.2 | |||
NM_016265.3 | 4154 | UTR 3 | NP_057349.2 |
Set Membership: |
HapMap |