Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613322 MIM: 613663 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GXYLT2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GXYLT2 - glucoside xylosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080393.1 | Intron | NP_001073862.1 | ||||
XM_011534067.2 | Intron | XP_011532369.1 | ||||
XM_011534068.1 | Intron | XP_011532370.1 | ||||
XM_017007123.1 | Intron | XP_016862612.1 | ||||
XM_017007124.1 | Intron | XP_016862613.1 |
SHQ1 - SHQ1, H/ACA ribonucleoprotein assembly factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |