Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCTTTGAGATGGGTTGTTCTTA[G/T]GTCACAGGACAGAGTGATCTGACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601582 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP5D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INPP5D - inositol polyphosphate-5-phosphatase D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017915.2 | Intron | NP_001017915.1 | ||||
NM_005541.4 | Intron | NP_005532.2 | ||||
XM_017004004.1 | Intron | XP_016859493.1 |