Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCAGTCTTGCTTGGCACTTGTCC[G/T]AATATGTGGTTAATTTAGCCTCTAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610801 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PM20D1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PM20D1 - peptidase M20 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152491.4 | Intron | NP_689704.4 | ||||
XM_017000396.1 | Intron | XP_016855885.1 | ||||
XM_017000397.1 | Intron | XP_016855886.1 | ||||
XM_017000398.1 | Intron | XP_016855887.1 |
SLC41A1 - solute carrier family 41 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |