Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTACCTTTATCATTGTCCTTCCAC[A/C]CCACTCATGTCCTTAATTCCTCCCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604496 MIM: 602916 | |||||||||||||||||||||||
Literature Links: |
NKIRAS1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
NKIRAS1 - NFKB inhibitor interacting Ras like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020345.3 | Intron | NP_065078.1 | ||||
XM_005265076.3 | Intron | XP_005265133.1 | ||||
XM_005265077.4 | Intron | XP_005265134.1 | ||||
XM_005265078.3 | Intron | XP_005265135.1 |
UBE2E1 - ubiquitin conjugating enzyme E2 E1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |