Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604756 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC441239 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC441239 - uncharacterized LOC441239 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011516745.2 | Intron | XP_011515047.1 |
ZNF273 - zinc finger protein 273 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021148.2 | Intron | NP_066971.2 | ||||
XM_011515746.2 | Intron | XP_011514048.1 | ||||
XM_017011688.1 | Intron | XP_016867177.1 | ||||
XM_017011689.1 | Intron | XP_016867178.1 | ||||
XM_017011690.1 | Intron | XP_016867179.1 | ||||
XM_017011691.1 | Intron | XP_016867180.1 | ||||
XM_017011692.1 | Intron | XP_016867181.1 | ||||
XM_017011693.1 | Intron | XP_016867182.1 | ||||
XM_017011694.1 | Intron | XP_016867183.1 | ||||
XM_017011695.1 | Intron | XP_016867184.1 | ||||
XM_017011696.1 | Intron | XP_016867185.1 | ||||
XM_017011697.1 | Intron | XP_016867186.1 | ||||
XM_017011698.1 | Intron | XP_016867187.1 |
Set Membership: |
HapMap |