Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAATTGATCCCATTTTGAATACA[A/G]AGATAGACCCCTTTTGAATATTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601058 MIM: 616375 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
H3F3B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
H3F3B - H3 histone, family 3B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4738 - microRNA 4738 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNK - unkempt family zinc finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080419.2 | Intron | NP_001073888.2 | ||||
XM_006722145.3 | Intron | XP_006722208.1 | ||||
XM_006722148.2 | Intron | XP_006722211.1 | ||||
XM_011525400.1 | Intron | XP_011523702.1 | ||||
XM_011525401.2 | Intron | XP_011523703.1 | ||||
XM_017025247.1 | Intron | XP_016880736.1 | ||||
XM_017025248.1 | Intron | XP_016880737.1 | ||||
XM_017025249.1 | Intron | XP_016880738.1 | ||||
XM_017025250.1 | Intron | XP_016880739.1 |