Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACCAATACAAAGTTAAAGGACA[A/T]GTCTCAAATTAGTAGAAGATATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186910 MIM: 186730 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD8A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD8A - CD8a molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CD8B - CD8b molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178100.1 | Intron | NP_001171571.1 | ||||
NM_004931.4 | Intron | NP_004922.1 | ||||
NM_172101.3 | Intron | NP_742099.1 | ||||
NM_172102.3 | Intron | NP_742100.1 | ||||
NM_172213.3 | Intron | NP_757362.1 | ||||
XM_011533164.2 | Intron | XP_011531466.1 |
Set Membership: |
HapMap |