Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615618 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POGLUT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
POGLUT1 - protein O-glucosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152305.2 | 357 | Silent Mutation | AGC,AGT | S,S 109 | NP_689518.1 | |
XM_006713705.3 | 357 | UTR 5 | XP_006713768.1 | |||
XM_017006878.1 | 357 | UTR 5 | XP_016862367.1 | |||
XM_017006879.1 | 357 | UTR 5 | XP_016862368.1 | |||
XM_017006880.1 | 357 | UTR 5 | XP_016862369.1 |