Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTACAAAGATACTGTGAAGAGA[C/T]AGTGCATGAAAATGTCAGAATACGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605357 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STON1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STON1 - stonin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198595.1 | Intron | NP_001185524.1 | ||||
NM_006873.3 | Intron | NP_006864.2 |
STON1-GTF2A1L - STON1-GTF2A1L readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198593.1 | Intron | NP_001185522.1 | ||||
NM_001198594.1 | Intron | NP_001185523.1 | ||||
NM_172311.2 | Intron | NP_758515.1 |
Set Membership: |
HapMap |