Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605119 MIM: 605963 MIM: 613598 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPM1G PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPM1G - protein phosphatase, Mg2+/Mn2+ dependent 1G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177983.2 | 2089 | UTR 3 | NP_817092.1 |
SNX17 - sorting nexin 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF513 - zinc finger protein 513 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201459.1 | 2089 | Intron | NP_001188388.1 | |||
NM_144631.5 | 2089 | Intron | NP_653232.3 | |||
XM_005264142.2 | 2089 | Intron | XP_005264199.1 | |||
XM_005264143.3 | 2089 | Intron | XP_005264200.1 |
Set Membership: |
HapMap |