Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAACGGTTCTGGTTTGGTTCCAG[C/T]GAATTTATGGGAAAGCAAAATAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182308 MIM: 604216 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC17A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC17A1 - solute carrier family 17 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005074.3 | Intron | NP_005065.2 | ||||
XM_011514818.2 | Intron | XP_011513120.2 | ||||
XM_011514819.2 | Intron | XP_011513121.2 | ||||
XM_011514820.2 | Intron | XP_011513122.2 | ||||
XM_011514821.2 | Intron | XP_011513123.1 | ||||
XM_017011199.1 | Intron | XP_016866688.1 | ||||
XM_017011200.1 | Intron | XP_016866689.1 | ||||
XM_017011201.1 | Intron | XP_016866690.1 | ||||
XM_017011202.1 | Intron | XP_016866691.1 |
SLC17A4 - solute carrier family 17 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286121.1 | Intron | NP_001273050.1 | ||||
NM_005495.2 | Intron | NP_005486.1 | ||||
XM_011514211.1 | Intron | XP_011512513.1 | ||||
XM_011514213.2 | Intron | XP_011512515.1 | ||||
XM_011514214.2 | Intron | XP_011512516.1 | ||||
XM_011514215.2 | Intron | XP_011512517.1 | ||||
XM_011514217.2 | Intron | XP_011512519.1 | ||||
XM_011514218.2 | Intron | XP_011512520.1 | ||||
XM_011514219.2 | Intron | XP_011512521.1 | ||||
XM_017010150.1 | Intron | XP_016865639.1 | ||||
XM_017010151.1 | Intron | XP_016865640.1 |
Set Membership: |
HapMap |