Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCACCTTTAGTTTATCCCTTCCC[A/C]CCTTTACTACATGCCACTTCATCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602745 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIP5K1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PIP5K1B - phosphatidylinositol-4-phosphate 5-kinase type 1 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278253.1 | Intron | NP_001265182.1 | ||||
NM_003558.3 | Intron | NP_003549.1 | ||||
XM_005252261.3 | Intron | XP_005252318.1 | ||||
XM_005252262.4 | Intron | XP_005252319.1 | ||||
XM_006717300.2 | Intron | XP_006717363.1 | ||||
XM_006717301.1 | Intron | XP_006717364.1 | ||||
XM_011519082.2 | Intron | XP_011517384.1 | ||||
XM_011519083.2 | Intron | XP_011517385.1 | ||||
XM_011519084.2 | Intron | XP_011517386.1 | ||||
XM_017015188.1 | Intron | XP_016870677.1 | ||||
XM_017015189.1 | Intron | XP_016870678.1 | ||||
XM_017015190.1 | Intron | XP_016870679.1 | ||||
XM_017015191.1 | Intron | XP_016870680.1 |
Set Membership: |
Validated |