Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAGTACAAGGAATTATGAGAAAT[A/T]GTTTGTTTGTGTTTTAATTGTTGTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600127 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE4B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PDE4B - phosphodiesterase 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037339.2 | Intron | NP_001032416.1 | ||||
NM_001037340.2 | Intron | NP_001032417.1 | ||||
NM_001037341.1 | Intron | NP_001032418.1 | ||||
NM_001297440.1 | Intron | NP_001284369.1 | ||||
NM_001297441.1 | Intron | NP_001284370.1 | ||||
NM_001297442.1 | Intron | NP_001284371.1 | ||||
NM_002600.3 | Intron | NP_002591.2 | ||||
XM_005270924.3 | Intron | XP_005270981.1 | ||||
XM_005270925.2 | Intron | XP_005270982.1 | ||||
XM_006710680.3 | Intron | XP_006710743.1 | ||||
XM_011541566.1 | Intron | XP_011539868.1 | ||||
XM_017001445.1 | Intron | XP_016856934.1 | ||||
XM_017001446.1 | Intron | XP_016856935.1 |
Set Membership: |
HapMap |