Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGCTGTCTGGGGGGCTGAGGGC[C/T]GGGCTGAGGCCTGTCCTGAGCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615921 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PERM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PERM1 - PPARGC1 and ESRR induced regulator, muscle 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291366.1 | 1289 | Missense Mutation | CAG,CGG | Q,R 524 | NP_001278295.1 | |
NM_001291367.1 | 1289 | Missense Mutation | CAG,CGG | Q,R 430 | NP_001278296.1 | |
XM_017002583.1 | 1289 | Missense Mutation | CAG,CGG | Q,R 524 | XP_016858072.1 | |
XM_017002584.1 | 1289 | Missense Mutation | CAG,CGG | Q,R 524 | XP_016858073.1 | |
XM_017002585.1 | 1289 | Missense Mutation | CAG,CGG | Q,R 524 | XP_016858074.1 |
PLEKHN1 - pleckstrin homology domain containing N1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |