Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608750 MIM: 608721 MIM: 610145 MIM: 611620 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALG3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ALG3 - ALG3, alpha-1,3- mannosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006941.2 | Intron | NP_001006942.1 | ||||
NM_005787.5 | Intron | NP_005778.1 | ||||
XM_011512322.1 | Intron | XP_011510624.1 | ||||
XM_011512323.2 | Intron | XP_011510625.1 |
CAMK2N2 - calcium/calmodulin dependent protein kinase II inhibitor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ECE2 - endothelin converting enzyme 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037324.2 | Intron | NP_001032401.1 | ||||
NM_001100120.1 | Intron | NP_001093590.1 | ||||
NM_001100121.1 | Intron | NP_001093591.1 | ||||
NM_014693.3 | Intron | NP_055508.3 | ||||
NM_032331.3 | Intron | NP_115707.2 |
MIR1224 - microRNA 1224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VWA5B2 - von Willebrand factor A domain containing 5B2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |