Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEIS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MEIS3 - Meis homeobox 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009813.2 | Intron | NP_001009813.1 | ||||
NM_001301059.1 | Intron | NP_001287988.1 | ||||
NM_020160.2 | Intron | NP_064545.1 | ||||
XM_011527136.2 | Intron | XP_011525438.1 | ||||
XM_011527138.2 | Intron | XP_011525440.1 | ||||
XM_011527139.2 | Intron | XP_011525441.1 | ||||
XM_011527140.1 | Intron | XP_011525442.1 | ||||
XM_017027011.1 | Intron | XP_016882500.1 |
Set Membership: |
HapMap Validated |