Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612183 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR176 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR176 - G protein-coupled receptor 176 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271854.1 | 6251 | Intron | NP_001258783.1 | |||
NM_001271855.1 | 6251 | Intron | NP_001258784.1 | |||
NM_007223.2 | 6251 | Intron | NP_009154.1 | |||
XM_011521167.2 | 6251 | UTR 3 | XP_011519469.1 | |||
XM_011521168.2 | 6251 | UTR 3 | XP_011519470.1 | |||
XM_017021873.1 | 6251 | UTR 3 | XP_016877362.1 | |||
XM_017021874.1 | 6251 | UTR 3 | XP_016877363.1 | |||
XM_017021875.1 | 6251 | UTR 3 | XP_016877364.1 | |||
XM_017021876.1 | 6251 | UTR 3 | XP_016877365.1 | |||
XM_017021877.1 | 6251 | UTR 3 | XP_016877366.1 | |||
XM_017021878.1 | 6251 | UTR 3 | XP_016877367.1 |