Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGACACAGGGGTCCCAGGCCCA[A/G]TGGCTCACTTTATCAGGGAAACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600856 MIM: 602631 MIM: 603240 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKN1C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CDKN1C - cyclin dependent kinase inhibitor 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A18 - solute carrier family 22 member 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A18AS - solute carrier family 22 member 18 antisense | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302862.1 | Intron | NP_001289791.1 | ||||
NM_007105.3 | Intron | NP_009036.2 | ||||
XM_017017832.1 | Intron | XP_016873321.1 | ||||
XM_017017833.1 | Intron | XP_016873322.1 | ||||
XM_017017834.1 | Intron | XP_016873323.1 | ||||
XM_017017835.1 | Intron | XP_016873324.1 | ||||
XM_017017836.1 | Intron | XP_016873325.1 |