Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTATGTGCCAACAGGCTGTAAG[A/G]GTTAGAAGTTAGTGTGTAATTTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609086 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL20 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FBXL20 - F-box and leucine rich repeat protein 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184906.1 | 9413 | UTR 3 | NP_001171835.1 | |||
NM_032875.2 | 9413 | UTR 3 | NP_116264.2 | |||
XM_005257746.3 | 9413 | UTR 3 | XP_005257803.2 | |||
XM_005257747.3 | 9413 | UTR 3 | XP_005257804.2 |
LOC101929578 - transcription factor CP2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |