Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAAAGAGGTAAAGTTATAGAAGA[A/T]TTAAAAAAGCCATGTGACTCTGTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609653 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTC-436P18.1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CTC-436P18.1 - uncharacterized LOC101928630 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFAF2 - NADH:ubiquinone oxidoreductase complex assembly factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMIM15 - small integral membrane protein 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001048249.3 | Intron | NP_001041714.1 |