Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120320 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL12A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
COL12A1 - collagen type XII alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004370.5 | 10149 | UTR 3 | NP_004361.3 | |||
NM_080645.2 | 10149 | UTR 3 | NP_542376.2 | |||
XM_011535434.1 | 10149 | Intron | XP_011533736.1 | |||
XM_011535435.1 | 10149 | Intron | XP_011533737.1 | |||
XM_011535436.2 | 10149 | Intron | XP_011533738.1 | |||
XM_017010252.1 | 10149 | Intron | XP_016865741.1 |
Set Membership: |
HapMap |