Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608300 MIM: 600781 | ||||||||||||||||||||
Literature Links: |
NAGS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAGS - N-acetylglutamate synthase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PYY - peptide YY | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004160.5 | 1421 | Intron | NP_004151.3 | |||
XM_011525035.1 | 1421 | Intron | XP_011523337.1 |
TMEM101 - transmembrane protein 101 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304813.1 | 1421 | UTR 3 | NP_001291742.1 | |||
NM_001304814.1 | 1421 | UTR 3 | NP_001291743.1 | |||
NM_032376.3 | 1421 | UTR 3 | NP_115752.1 | |||
XM_005257737.2 | 1421 | UTR 3 | XP_005257794.1 | |||
XM_011525346.1 | 1421 | UTR 3 | XP_011523648.1 | |||
XM_011525347.1 | 1421 | UTR 3 | XP_011523649.1 | |||
XM_011525349.1 | 1421 | UTR 3 | XP_011523651.1 | |||
XM_011525351.1 | 1421 | UTR 3 | XP_011523653.1 | |||
XM_011525353.1 | 1421 | UTR 3 | XP_011523655.1 |