Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGAGCTACAGGCCCTGGAGGCCAT[C/T]TACGGCGCGGACTTCCAAGACCTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609280 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF2AK4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
EIF2AK4 - eukaryotic translation initiation factor 2 alpha kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013703.3 | 194 | Silent Mutation | ATC,ATT | I,I 33 | NP_001013725.2 | |
XM_005254392.2 | 194 | Silent Mutation | ATC,ATT | I,I 33 | XP_005254449.1 | |
XM_011521599.2 | 194 | Silent Mutation | ATC,ATT | I,I 33 | XP_011519901.1 | |
XM_011521600.2 | 194 | Silent Mutation | ATC,ATT | I,I 33 | XP_011519902.1 | |
XM_017022219.1 | 194 | Silent Mutation | ATC,ATT | I,I 33 | XP_016877708.1 |
LOC100505534 - uncharacterized LOC100505534 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |