Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCEAL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TCEAL4 - transcription elongation factor A like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006935.2 | 9 | UTR 5 | NP_001006936.1 | |||
NM_001006937.2 | 9 | UTR 5 | NP_001006938.1 | |||
NM_001300901.1 | 9 | Intron | NP_001287830.1 | |||
NM_001305840.1 | 9 | UTR 5 | NP_001292769.1 | |||
NM_001305841.1 | 9 | UTR 5 | NP_001292770.1 | |||
NM_001305842.1 | 9 | UTR 5 | NP_001292771.1 | |||
NM_024863.5 | 9 | UTR 5 | NP_079139.4 | |||
XM_017029856.1 | 9 | Intron | XP_016885345.1 | |||
XM_017029857.1 | 9 | Intron | XP_016885346.1 |
Set Membership: |
HapMap Validated |