Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCTTTTTCTCAGTGTTCTATGT[A/G]TCAAGCCTGATGGGAAATCTCCTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371030 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC105371030 - uncharacterized LOC105371030 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR4F6 - olfactory receptor family 4 subfamily F member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005326.1 | 108 | Silent Mutation | GTA,GTG | V,V 36 | NP_001005326.1 |
Set Membership: |
HapMap |