Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300608 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DACH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DACH2 - dachshund family transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139514.1 | Intron | NP_001132986.1 | ||||
NM_001139515.1 | Intron | NP_001132987.1 | ||||
NM_053281.3 | Intron | NP_444511.1 | ||||
XM_011530846.2 | Intron | XP_011529148.1 | ||||
XM_011530847.2 | Intron | XP_011529149.1 | ||||
XM_011530848.2 | Intron | XP_011529150.1 | ||||
XM_017029254.1 | Intron | XP_016884743.1 | ||||
XM_017029255.1 | Intron | XP_016884744.1 | ||||
XM_017029256.1 | Intron | XP_016884745.1 |
Set Membership: |
HapMap |